Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-PIK3C3 (Ser164) Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS5581R
Description
VPS34 (PIK3C3) is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate; different complex forms are believed to play a role in multiple membrane trafficking pathways: PI3KC3-C1 is involved in initiation of autophagosomes and PI3KC3-C2 in maturation of autophagosomes and endocytosis. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.Specifications
Phospho-PIK3C3 (Ser164) | |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Pik3c3 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
5289 |
Pik3c3 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8NEB9 | |
5330434F23Rik; catalytic phosphatidylinositol 3-kinase 3; catalytic subunit type 3 Antibody; class 3 phosphoinositide-3-kinase; Class III PI 3-kinase; hVps34; hVps34 Antibody; OTTMUSP00000023341; OTTMUSP00000023350; phosphatidylinositol 3-kinase; Phosphatidylinositol 3-kinase catalytic subunit type 3; Phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI3K type 3; PI-3K Vps34p; PI3-kinase type 3; Pik3c3; PIK3C3 Antibody; PtdIns-3-kinase type 3; Vps34 | |
KLH conjugated synthetic phosphopeptide derived from human PIK3C3 around the phosphorylation site of Ser164. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title