Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PKC beta 1 (Ser642) Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS5566R

Catalog No. 50-198-6653


Add to cart

Description

Description

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

Phospho-PKC beta 1 (Ser642)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PRKCB
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
5579
PRKCB
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P05771
A130082F03Rik; PKCB; PKC-B; PKC-beta; Prkcb; PRKCB1; Prkcb2; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; Protein kinase C beta type; protein kinase C beta-II; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide
KLH conjugated synthetic phosphopeptide derived from human PKC beta 1 around the phosphorylation site of Ser642.
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit