Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Phospho-PLK1 (Ser326) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 195531AP
Catalog No. 50-173-2452
Description
Description
The antibody is specific to Ser(326)-phospho.
PLK1 (Polo Like Kinase 1) is a Protein Coding gene. Diseases associated with PLK1 include Transient Tic Disorder and Monoclonal Paraproteinemia. Among its related pathways are Integrated Breast Cancer Pathway and Cell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is PLK3. This gene encodes a protein which contains a conserved nuclear localization signal.
Specifications
Specifications
Phospho-PLK1 (Ser326)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53350, Q07832, Q62673
PLK, PLK 1, PLK1, Polo like kinase 1, STPK13
Rabbit
IgG
150 μL
-20°C
Primary
18817, 25515, 5347
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PLK1
Liquid
Plk1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat