Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-PPARgamma (Ser112) Rabbit anti-Mouse,Canine,Human,Rat, KHL, Clone: AW504, MilliporeSigma™

Rabbit Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen phospho-PPARgamma (Ser112)
Applications Western Blot
Conjugate KHL
Format Culture Supernatant
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
04816I100MI
View Documents
MilliporeSigma
04816I100UL
100 μL
Each for $386.00
Add to cart
 
04816I25UMI
View Documents
MilliporeSigma
04816I25UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

phospho-PPARgamma (Ser112) Monoclonal antibody specifically detects phospho-PPARgamma (Ser112) in Mouse,Canine,Human,Rat samples. It is validated for Western Blotting.
Specifications

Specifications

phospho-PPARgamma (Ser112)
KHL
Rabbit
Peroxisome proliferator-activated receptor gamma;Nuclear receptor subfamily 1 group C member 3
KLH-conjugated linear peptide corresponding to 12 amino acids surrounding phosphoserine 112 from human PPAR gamma, isoform 2.
Primary
Stable for 1 year at -20°C from date of receipt.
Western Blot
Culture Supernatant
Mouse, Canine, Human, Rat
PPARG, NR1C3
Monoclonal
Unpurified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit