Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PPIG (Ser376) Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS3461R

Catalog No. 50-198-5422


Add to cart

Description

Description

Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregation defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
Specifications

Specifications

Phospho-PPIG (Ser376)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PPIG
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
9360
PPIG
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q13427
AU019516; AU022200; B230312B02Rik; BOS_1662; CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; CYP; LOC100341939; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); Matrin-cyp; MGC133241; OTTHUMP00000204761; OTTHUMP00000204762; Peptidyl-prolyl cis-trans isomerase G; peptidylprolyl isomerase G; peptidyl-prolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); peptidyl-prolyl isomerase G (cyclophilin G); peptidylprolyl isomerase G-like; PPIase G; PPIG; Rotamase G; SCAF10; SR-cyclophilin; SRCyp; SR-cyp; SR-related CTD-associated factor 10
KLH conjugated synthetic phosphopeptide derived from human PPIG around the phosphorylation site of Ser376 [VS (p-S)GE].
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit