Learn More
Phospho-PRKD3 (Ser41) Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS5564R
Catalog No. 50-198-6652
Add to cart
Description
Description
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role. The protein encoded by this gene is one of the PKC family members. This kinase can be activated rapidly by the agonists of G protein-coupled receptors. It resides in both cytoplasm and nucleus, and its nuclear accumulation is found to be dramatically enhanced in response to its activation. This kinase can also be activated after B-cell antigen receptor (BCR) engagement, which requires intact phopholipase C gamma and the involvement of other PKC family members.
Specifications
Specifications
Phospho-PRKD3 (Ser41)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Prkd3
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
23683
Prkd3
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
O94806
4930557O20Rik; 5730497N19Rik; EPK2; nPKC-nu; Pkcnu; PKC-NU; PKD3; Prkcn; Prkd3; Protein kinase C nu type; protein kinase C, nu; protein kinase D3; Protein kinase EPK2; Serine/threonine-protein kinase D3
KLH conjugated synthetic phosphopeptide derived from human PRKD3 around the phosphorylation site of Ser41 [RL (p-S)NG].
Protein A
RUO
Antibody
Polyclonal
Human