Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-RSK3 (S218) Rabbit anti-Human, Mouse, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Supplier: R&D Systems AF893SP
This item is not returnable.
View return policy
Description
Phospho-RSK3 (S218) Polyclonal antibody AF893SP specifically detects Phospho-RSK3 (S218) in Human, Mouse samples. It is validated for Immunohistochemistry, Western BlottingSpecifications
RSK3 (p ser218) | |
Polyclonal | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
EC 2.7.11, EC 2.7.11.1, HU-2, MAP kinase-activated protein kinase 1c, MAPK-activated protein kinase 1c, MAPKAP kinase 1c, MAPKAPK-1c, MAPKAPK1C, p90-RSK 2, p90RSK2, p90-RSK3, ribosomal protein S6 kinase alpha 2, ribosomal protein S6 kinase alpha-2, ribosomal protein S6 kinase, 90kD, polypeptide 2, ribosomal protein S6 kinase, 90kDa, polypeptide 2, Ribosomal S6 kinase 3,90 kDa ribosomal protein S6 kinase 2, RPS6KA2, RSK, RSK-3, RSK3pp90RSK3, S6K-alpha, S6K-alpha2, S6K-alpha-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
6196 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
RPS6KA2 | |
Phosphopeptide containing human and mouse RSK3 S218 site | |
25 μg | |
Primary | |
Detects human and mouse RSK3 when phosphorylated at S218. Reactivity with other RSK isoforms when phosphorylated at corresponding sites is unknown. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction