Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SF1 (Ser82) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540282

Catalog No. PA540282


Add to cart

Description

Description

This antibody is predicted to react with mouse and monkey based on sequence homology.

Necessary for the ATP-dependent first step of spliceosome assembly. Binds to the intron branch point sequence (BPS) 5 ft.-UACUAAC-3 ft. of the pre-mRNA. May act as transcription repressor.
Specifications

Specifications

Phospho-SF1 (Ser82)
Polyclonal
Unconjugated
SF1
Liquid
SF1
Synthesized peptide derived from human Splicing factor 1 around the phosphorylation site of S82.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q15637
SF1; ZFM1; ZNF162; Splicing factor 1; Mammalian branch point-binding protein; BBP; mBBP; Transcription factor ZFM1; Zinc finger gene in MEN1 locus; Zinc finger protein 162
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7536
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit