Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGK223 Polyclonal Antibody
Phospho-SGK223 (Tyr413) Polyclonal Antibody
Supplier: Thermo Scientific PA5103157
Description
Phospho-SGK223 (Tyr413) Polyclonal Antibody for Western Blot

Specifications
SGK223 | |
Polyclonal | |
Unconjugated | |
PRAG1 | |
9830148H23; D8Ertd82e; DNA segment, Chr 8, ERATO Doi 82, expressed; homolog of rat pragma of Rnd2; Inactive tyrosine-protein kinase PRAG1; mFLJ00269; NACK; Notch activation complex kinase; PEAK1-related kinase-activating pseudokinase 1; Prag1; Pragma of Rnd2; pragmin; RGD1311793; Sgk223; Sugen kinase 223; tyrosine-protein kinase SgK223 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
157285, 244418, 306506 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q571I4, Q86YV5 | |
PRAG1 | |
A synthesized peptide derived from human PRAG1(Accession Q86YV5), corresponding to amino acid residues I1047-H1097. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction