Learn More
phospho-Smad2 (Ser465/467) Rabbit anti-Human,Mouse,Rat, KHL, Clone: A5S, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB0495325UL
 View more versions of this product
Catalog No. ZRB049532MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
phospho-Smad2 (Ser465/467) Monoclonal antibody specifically detects phospho-Smad2 (Ser465/467) in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Inhibition Assay, Western Blotting.
Specifications
Specifications
phospho-Smad2 (Ser465/467)
A5S
KHL
Mothers against decapentaplegic homolog 2;MAD homolog 2;Mothers against DPP homolog 2;JV18-1;Mad-related protein 2;hMAD-2;SMAD family member 2;hSMAD2
Rabbit
IgG
Protein A Purified
Signaling
Primary
Human, Mouse, Rat
Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
SMAD2, MADH2, MADR2
KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 465 and 467 from human Smad2.
∼55 kDa observed; 52.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal