Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-Smad2 (Ser465/467) Rabbit anti-Human,Mouse,Rat, KHL, Clone: A5S, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen phospho-Smad2 (Ser465/467)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB049532MI
View Documents
MilliporeSigma
ZRB0495325UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB049534MI
View Documents
MilliporeSigma
ZRB049534X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

phospho-Smad2 (Ser465/467) Monoclonal antibody specifically detects phospho-Smad2 (Ser465/467) in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

phospho-Smad2 (Ser465/467)
KHL
Rabbit
Mothers against decapentaplegic homolog 2;MAD homolog 2;Mothers against DPP homolog 2;JV18-1;Mad-related protein 2;hMAD-2;SMAD family member 2;hSMAD2
KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 465 and 467 from human Smad2.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Purified
Human, Mouse, Rat
SMAD2, MADH2, MADR2
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit