Learn More
Phospho-Smtnl1 (Ser301) Rabbit anti-Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 195601AP
Catalog No. 50-173-2455
Add to cart
Description
Description
The antibody recognize Ser(301) phospho of Smtn1.
Plays a role in the regulation of contractile properties of both striated and smooth muscles. When unphosphorylated, may inhibit myosin dephosphorylation. Phosphorylation at Ser-299 reduces this inhibitory activity.
Specifications
Specifications
Phospho-Smtnl1 (Ser301)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99LM3
1110030K22Rik, CHASM, smoothelin like 1, Smtnl1
Rabbit
IgG
150 μL
-20°C
Primary
68678
Western Blot
Unconjugated
Smtnl1
Liquid
Smtnl1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse