Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-STAT1 (Ser727) Rabbit anti-Human, Mouse, Rat, Clone: SN67-04, Invitrogen™

Rabbit Monoclonal Antibody

Manufacturer:  Thermo Scientific MA532271

Catalog No. PIMA532271


Add to cart

Description

Description

Rabbit Monoclonal Antibody

Phospho-STAT1 (Ser727) Monoclonal antibody specifically detects Phospho-STAT1 (Ser727) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot
Specifications

Specifications

Phospho-STAT1 (Ser727)
Monoclonal
1 mg/mL
TBS with 1% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
P42224, P42225
2010005J02Rik; AA408197; BOS_1917; CANDF7; DD6G4-4; DKFZp686B04100; IMD31A; IMD31B; IMD31C; ISGF-3; OTTHUMP00000165046; OTTHUMP00000205845; Signal transducer and activator of transcription 1; Signal transducer and activator of transcription 1-alpha/beta; STAT; STAT1; STAT91; Transcription factor ISGF-3 components p91/p84
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20846, 25124, 6772
Immunohistochemistry, Western Blot
SN67-04
Unconjugated
STAT1
Liquid
STAT1
Synthetic phospho-peptide corresponding to residues surrounding Ser727 of human STAT1
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit