Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-Stat3 (Tyr705) Rabbit anti-Human,Monkey,Mouse,Rat,Bovine, KHL, Clone: 6C3, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen phospho-Stat3 (Tyr705)
Applications Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB100625MI
View Documents
MilliporeSigma
ZRB100625UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10064XMI
View Documents
MilliporeSigma
ZRB10064X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

phospho-Stat3 (Tyr705) Monoclonal antibody specifically detects phospho-Stat3 (Tyr705) in Human,Monkey,Mouse,Rat,Bovine samples. It is validated for Immunohistochemistry (Paraffin), Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

phospho-Stat3 (Tyr705)
KHL
Rabbit
Signal transducer and activator of transcription 3;Acute-phase response factor
KLH-conjugated linear peptide corresponding to 11 amino acids from the C-terminal region of human Stat3 surrounding phospho tyrosine 705.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Purified
Human, Monkey, Mouse, Rat, Bovine
STAT3, APRF
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit