Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Stat5 (Tyr694) Recombinant Rabbit Monoclonal Antibody (Stat5Y694-G11), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$206.50 - $507.00
Specifications
Antigen | Phospho-Stat5 (Tyr694) |
---|---|
Clone | Stat5Y694-G11 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA537356
|
Thermo Scientific
MA537356 |
10 Tests |
Each for $206.50
|
|
|||||
PIMA537110
|
Thermo Scientific
MA537110 |
100 Tests |
Each for $507.00
|
|
|||||
Description
Phospho-Stat5 (Tyr694) Recombinant Monoclonal antibody specifically detects Phospho-Stat5 (Tyr694) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Phospho-Stat5 (Tyr694) | |
Flow Cytometry | |
FITC | |
Rabbit | |
Human, Mouse | |
P42229, P42230 | |
20850, 6776 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr694 of human phospho Stat5. | |
Primary | |
4° C, store in dark | |
STAT5A |
Stat5Y694-G11 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5 | |
STAT5A | |
IgG κ | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title