Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STAT5A/B (Tyr694, Tyr699) Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
$147.50 - $565.00
Specifications
Antigen | Phospho-STAT5A/B (Tyr694, Tyr699) |
---|---|
Concentration | Lot-Specific |
Applications | Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2738
|
Active Motif
39618 |
10 μL |
Each for $147.50
|
|
|||||
50-199-2737
|
Active Motif
39617 |
200 μL |
Each for $565.00
|
|
|||||
Description
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor-associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated by, and mediates the responses of, many cell ligands, such as IL-2, IL-3, IL-7, GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of this protein in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of this gene is found to induce the expression of BCL2L1/ BCL-X(L), which suggests the antiapoptotic function of this gene in cells.Specifications
Phospho-STAT5A/B (Tyr694, Tyr699) | |
Dot Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P42229, P51692 | |
6776, 6777 | |
This STAT5A/B phospho Tyr694/Tyr699 antibody was raised against a peptide containing phospho Tyr694 of human STAT5A. Human STAT5B shares the identical sequence surrounding Tyr699. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
STAT5B |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
AA959963; Mammary gland factor; mammary gland factor STAT5A; mammary gland factor STAT5B; Mgf; Mpf; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5A; Signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; Stat5a; STAT5B; transcription factor STAT5B | |
STAT5A, STAT5B | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title