Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SYN1 (Ser62, Ser67) Rabbit anti-Human|Mouse|Rat, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS5632R

Catalog No. 50-198-6692


Add to cart

Description

Description

SYN1 (synapsin 1) is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins that associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains and are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. Synapsin 1 plays a role in regulation of axonogenesis and synaptogenesis. It serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

Phospho-SYN1 (Ser62, Ser67)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SYN1
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
20964, 24949, 6853
SYN1
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
O88935, P09951, P17600
BOS_25913; Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; Synapsin I; Synapsin-1; SYNI; truncated synapsin 1-S
KLH conjugated synthetic phosphopeptide derived from human SYN1 around the phosphorylation site of Ser62+Ser67.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit