Learn More
Phospho-TPH2 (Ser19) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA14736
Catalog No. PIPA14736
Add to Cart
Description
Description
This antibody is predicted to react with bovine, mouse and zebrafish based on 100% sequence homology.
Phospho-TPH2 (Ser19) Polyclonal antibody specifically detects Phospho-TPH2 (Ser19) in Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-TPH2 (Ser19)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q8CGU9, Q8CGV2
Tph2
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser19 of rat TPH2 conjugated to KLH
100 μL
Primary
Mouse, Rat
Antibody
IgG
Western Blot
Unconjugated
Tph2
ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2
Rabbit
Affinity chromatography
RUO
216343, 317675
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Recommendations
Recommendations