Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tyrosine Rabbit anti-All, Clone: PY2870R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Phospho-Tyrosine
Clone PY2870R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122050
View Documents
Novus Biologicals
NBP30728820UG
20 μg
Each for $159.82
Add to cart
 
NB122049
View Documents
Novus Biologicals
NBP30728810UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in All samples. It is validated for ELISA
Specifications

Specifications

Phospho-Tyrosine
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
Recombinant full-length human Phospho-Tyrosine protein
Monoclonal
Protein A or G purified
PY2870R
ELISA
Unconjugated
Rabbit
All
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit