Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Wee1 (Ser642) Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS3474R
Description
Wee1 is a nuclear protein, which is a tyrosine kinase belonging to the Ser/Thr family of protein kinases. This protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase.Specifications
Phospho-Wee1 (Ser642) | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
WEE1 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
7465 |
WEE1 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P30291 | |
BOS_14491; DKFZp686I18166; fa20h03; FLJ16446; wee 1 homolog; WEE 1 homolog 1 (S. pombe); WEE1; WEE1 G2 checkpoint kinase; WEE1 homolog; WEE1 homolog (S. pombe); wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; Wee1A kinase; WEE1hu; Wee1-like protein kinase; wu:fa20h03; zgc:136514 | |
KLH conjugated synthetic phosphopeptide derived from human Wee1 around the phosphorylation site of Ser642 [SV (p-S)LT]. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title