Learn More
Phospho-ZAP70/Syk (Tyr493, Tyr526) Rabbit anti-Human, Mouse, FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Manufacturer:  Invitrogen MA528071
Catalog No. PIMA528071
Add to cart
Description
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using Recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
ZAP-70 and SYK are members of the SYK protein tyrosine kinase (PTK) family that are phosphorylated and activated by Src family PTKs. ZAP-70/SYK Y319/Y352 are located in the so-called interdomain of ZAP-70/SYK (between the N-terminal dual SH2 domains and the C-terminal kinase domain). Phosphorylation of ZAP-70 Y319 by Lck is necessary for T cell receptor (TCR)-dependent association of ZAP-70 with Lck and phospholipase C gamma and subsequent activation of calcium-dependent and Ras signaling cascades. SYK Y352 phosphorylation by Fyn/Lyn is critical for propagation of B cell receptor (BCR) signaling and for B cell development.
Specifications
Specifications
Phospho-ZAP70/Syk (Tyr493, Tyr526)
Recombinant Monoclonal
FITC
ZAP70
Liquid
Rabbit
IgG1, kappa
100 tests
4° C
Primary
20963, 22637, 6850, 7535
Flow Cytometry
Zap70Y493-H11
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P43403, P43404, P43405, P48025
SYK, ZAP70
A synthetic phospho-peptide corresponding to residues surrounding Tyr493 of human phospho Zap70
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse