Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHOSPHO1 Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300091B

Catalog No. NB064374


Add to cart

Description

Description

PHOSPHO1 Polyclonal antibody specifically detects PHOSPHO1 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

PHOSPHO1
Biotin
PBS
EC 3.1.3.75, phosphatase, orphan 1, phosphoethanolamine/phosphocholine phosphatase
Produced in rabbits immunized with purified, recombinant PHOSPHO1 (Q8TCT1-1; Met1-Cys267).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Protein Phosphatase
Primary
162466
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit