Learn More
Alfa Aesar™ Phosphodiesterase 4 Inhibitor
Manufacturer:  Alfa Aesar™ J64453LB0
 View more versions of this product
Catalog No. AAJ64453LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD.
Specifications
Specifications
5mg
2° to 8°C
Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD
C14H15N3O4
Ethyl 3,5-dimethyl-1-(3-nitrophenyl)-1H-pyrazole-4-carboxylate
Signal Transduction Reagent
Pale Yellow
Solid
346440-86-6
289.29g/mol
Light Sensitive