Alfa Aesar™ Phosphoenolpyruvic acid mono(cyclohexylammonium) salt, 98%

Manufacturer: Alfa Aesar™ A1238503

Quantity 1g

 View more versions of this product

Catalog No. AAA1238503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10526-80-4
    Molecular Formula C9H18NO6P
    Formula Weight 267.22
    MDL Number MFCD00036375
    Synonym PEP-CHA
    Chemical Name or Material Phosphoenolpyruvic acid mono (cyclohexylammonium) salt

    Specifications

    Formula Weight 267.22
    TSCA TSCA
    Molecular Formula C9H18NO6P
    CAS 10526-80-4
    Synonym PEP-CHA
    MDL Number MFCD00036375
    Recommended Storage −30°C to −10°C
    Melting Point ≈140°C (dec.)
    Quantity 1g
    Chemical Name or Material Phosphoenolpyruvic acid mono (cyclohexylammonium) salt
    Assay 98%

    −30°C to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit