Learn More
Phospholamban Rabbit anti-Human,Mouse,Rat, KHL, Clone: 4J16, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB121225UL
 View more versions of this product
Catalog No. ZRB121225MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Phospholamban Monoclonal antibody specifically detects Phospholamban in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Phospholamban
4J16
KHL
Purified
PLN, PLB
KLH-conjugated linear peptide corresponding to 23 amino acids from human Cardiac phospholamban.
Target Molecular Weight 6.11 kDa calculated; ∼6.0 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_002658.1
Cardiac phospholamban;PLB
Rabbit
IgG
Protein A Purified
Epigenetics and Nuclear Function
Primary
Human, Mouse, Rat