Learn More
Phospholipase A2 XII Mouse anti-Human, FITC, Clone: 3H2C11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237255F
Catalog No. NBP237255F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Phospholipase A2 XII
3H2C11
Flow Cytometry, ELISA, Immunohistochemistry
Purified
PLA2G12A
Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. Coli.
Protein A or G purified
Lipid and Metabolism
Primary
81579
ELISA, Flow Cytometry, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
EC 3.1.1.4, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, GXII, GXII sPLA2, Phosphatidylcholine 2-acylhydrolase 12A, phospholipase A2, group XII, phospholipase A2, group XIIA, PLA2G12, ROSSY, sPLA2-XII
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human