Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospholipase A2 XII Mouse anti-Human, PE, Clone: 7C7C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237258PE

Catalog No. NBP237258PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Phospholipase A2 XII
7C7C9
Flow Cytometry
Purified
PLA2G12A
Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. Coli.
Protein A or G purified
Lipid and Metabolism
Primary
81579
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
EC 3.1.1.4, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, GXII, GXII sPLA2, Phosphatidylcholine 2-acylhydrolase 12A, phospholipase A2, group XII, phospholipase A2, group XIIA, PLA2G12, ROSSY, sPLA2-XII
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit