Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipase B1 Mouse anti-Human, DyLight™ 650, Clone: OTI1D3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273386C
Description
Phospholipase B1 Monoclonal specifically detects Phospholipase B1 in Human samples. It is validated for Western Blot.Specifications
Phospholipase B1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLB1 | |
Full length recombinant protein of human Phospholipase B1 (NP_694566) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI1D3 | |
Western Blot | |
FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
151056.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title