Learn More
Medchemexpress LLC Phospholipase D, Streptomyces chromofuscus (PLD) | 9001-87-0 | 5 KU
SDP
Supplier:  Medchemexpress LLC HYP28125KU
Encompass_Preferred
Phospholipase D, Streptomyces chromofuscus (PLD) is an enzyme of the phospholipase superfamily, found across various life forms and frequently used in biochemical research. It catalyzes the hydrolysis of phosphodiester bonds in glycerophospholipids, yielding phosphatidic acid and soluble choline. This enzyme is implicated in various disease-related processes. This product is an enzyme preparation derived from natural strains through deep liquid fermentation, containing highly active phospholipase.
  • Catalyzes hydrolysis of phosphodiester bonds in glycerophospholipids.
  • Produces phosphatidic acid and choline.
  • Specifically catalyzes hydrolysis of phosphatidyl diester bond at the end of phosphatidylcholine.
  • Enzyme activity is ≥50000 U/mL.
  • Effective temperature range 35°C-55°C, optimum 40°C-50°C.
  • Effective pH range pH4.0-7.5, optimum pH 5.0-6.0.
  • Soluble in H2O at ≥ 100 mg/mL.
Catalog No. 50-003-99468