Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic acid hydrate, 80%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
51429-74-4, 12026-57-2
H3Mo12O40P·xH2O
1825.25
MFCD00149913
Molybdophosphoric acid
51429-74-4, 12026-57-2
H3Mo12O40P·xH2O
1825.25
MFCD00149913
Molybdophosphoric acid
Oxidation reagent
Chloride 200ppm max
Ammonium 100ppm max
Sulfate 250ppm max
Quantity 500g
Packaging Glass bottle
Color Yellow
Loss on Drying 25% max.
Assay Percent Range 47.5% min. (Mo)
Linear Formula H3[P(Mo3O10)4]·xH2O
Merck Index 14, 7342
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 1825.25
Physical Form Crystals
Percent Purity 80%
Chemical Name or Material Phosphomolybdic acid hydrate, ca. 78% Phosphomolybdic Acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger