Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic Acid, x-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

51429-74-4
H3PMo12O40·xH2O

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
CAS 51429-74-4
Molecular Formula H3PMo12O40·xH2O
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.10kg
Chemical Name or Material Phosphomolybdic Acid, x-Hydrate
Grade Baker Analyzed™ ACS Reagent