Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphonitrilic chloride trimer, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
940-71-6
Cl6N3P3
347.65
MFCD00006474
UBIJTWDKTYCPMQ-UHFFFAOYSA-N
phosphonitrilic chloride trimer,hexachlorocyclotriphosphazene,hexachlorophosphazene,triphosphonitrilic chloride,triphosphonitrile chloride,hexachlorotriphosphonitrile,cyclophosphazene dichloride trimer,hexachlorocyclophosphazatriene,hexachlorocyclotriphosphazatriene,phosphononitrilic chloride trimer
220225
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl
940-71-6
Cl6N3P3
347.65
MFCD00006474
UBIJTWDKTYCPMQ-UHFFFAOYSA-N
phosphonitrilic chloride trimer,hexachlorocyclotriphosphazene,hexachlorophosphazene,triphosphonitrilic chloride,triphosphonitrile chloride,hexachlorotriphosphonitrile,cyclophosphazene dichloride trimer,hexachlorocyclophosphazatriene,hexachlorocyclotriphosphazatriene,phosphononitrilic chloride trimer
220225
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl
Assay Percent Range 98%
Fieser 02,206; 04,386; 06,469
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether,benzene,toluene
Formula Weight 347.65
Percent Purity 98%
Density 1.9800g/mL
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in ether, benzene, toluene
Specific Gravity 1.98
Melting Point 111.0°C to 115.0°C
Quantity 100g
Chemical Name or Material Phosphonitrilic chloride trimer
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 213-376-8