Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphonitrilic Chloride Trimer 98.0+%, TCI America™

Click to view available options
Quantity:
25 g
250 g
940-71-6
Cl6N3P3
347.642
MFCD00006474
UBIJTWDKTYCPMQ-UHFFFAOYSA-N
phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
220225
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl
940-71-6
Cl6N3P3
347.642
MFCD00006474
UBIJTWDKTYCPMQ-UHFFFAOYSA-N
phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
220225
2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl
Melting Point 113°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
UN Number 1759
Formula Weight 347.64
Percent Purity ≥98.0% (GC)
Chemical Name or Material Phosphonitrilic Chloride Trimer

EINECSNumber : (1)-0652

TSCA : Yes