Alfa Aesar™ Phosphonoacetic acid, 98+%

Manufacturer: Alfa Aesar™ A1211722

Quantity 100g

 View more versions of this product

Catalog No. AAA1211722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4408-78-0
    Molecular Formula C2H5O5P
    Formula Weight 140.04
    MDL Number MFCD00004311
    UN Number UN3261
    Beilstein 1764355
    Chemical Name or Material Phosphonoacetic acid

    Specifications

    Formula Weight 140.04
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    MDL Number MFCD00004311
    Beilstein 1764355
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    RTECS Number AJ3278000
    CAS 4408-78-0
    UN Number UN3261
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Odor Odorless
    Quantity 100g
    Molecular Formula C2H5O5P
    Melting Point 139° to 146°C
    Chemical Name or Material Phosphonoacetic acid
    Assay ≥98%

    Description

    Applications
    Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit