Learn More
Phosphonoacetic acid, 98+%, Thermo Scientific Chemicals
Description
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 139°C to 146°C |
Odor | Odorless |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 1764355 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 140.04 |
Percent Purity | ≥98% |
Chemical Name or Material | Phosphonoacetic acid |
Safety and Handling
Hazard Category | Skin corrosion/irritation (category 1) |
Hazard Statement | Causes severe skin burns and eye damage |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 224-558-1 |
RTECSNumber | AJ3278000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.