Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphopantothenate-cysteine ligase Mouse anti-Human, Clone: 10D11, Novus Biologicals™
Mouse Monoclonal Antibody
$337.00 - $482.50
Specifications
Antigen | Phosphopantothenate-cysteine ligase |
---|---|
Clone | 10D11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1078, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25948150
|
Novus Biologicals
NBP25948150UL |
50 μL |
Each for $337.00
|
|
|||||
NBP259481X
|
Novus Biologicals
NBP259481100UL |
100 μL |
Each for $482.50
|
|
|||||
Description
Phosphopantothenate-cysteine ligase Monoclonal specifically detects Phosphopantothenate-cysteine ligase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Phosphopantothenate-cysteine ligase | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
FLJ11838, MGC117357, phosphopantothenoylcysteine synthetase, PPC synthetase | |
PPCS | |
IgG1 κ | |
Protein A purified |
10D11 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1078, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
79717 | |
Recombinant human PPCS (1-311aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title