Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 10% (v/v) Aqueous Solution, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Drum
7664-38-2, 7732-18-5
7664-38-2, 7732-18-5
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.