Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoric Acid, 25% (v/v), Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Color | Colorless |
Concentration | 25% (v/v) |
CAS Min % | 74.0, 24.0 |
CAS Max % | 76.0, 26.0 |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Quantity | 500 mL |
Physical Form | Liquid |
Percent Purity | 25% V/V |
Grade | Laboratory |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction