Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 50% v/v, Ricca Chemical

Click to view available options
Quantity:
4L
Packaging:
Poly natural
7664-38-2,7732-18-5
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
orthophosphoric acid,sonac,o-phosphoric acid,phosphorsaeure,acidum phosphoricum,evits,wc-reiniger,acide phosphorique,white,acido fosforico
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
7664-38-2,7732-18-5
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
orthophosphoric acid,sonac,o-phosphoric acid,phosphorsaeure,acidum phosphoricum,evits,wc-reiniger,acide phosphorique,white,acido fosforico
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
Color Colorless
Quantity 4L
CAS Min % 49.0,49.0
CAS Max % 51.0,51.0
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 50% v/v
Chemical Name or Material Phosphoric Acid