Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
PVC Coated Glass Bottle

Chemical Identifiers

7664-38-2
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Formula Weight 98
Packaging PVC Coated Glass Bottle
Specific Gravity 1L = 1.69kg
Chemical Name or Material Phosphoric Acid
Grade Baker Analyzed™ ACS Reagent