Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5L
500mL
Packaging:
Glass Bottle
PVC Coated Glass Bottle

Chemical Identifiers

7664-38-2
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
CHEBI:26078

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES OP(=O)(O)O
IUPAC Name phosphoric acid
Molecular Weight (g/mol) 97.994
PubChem CID 1004
ChEBI CHEBI:26078
Formula Weight 98
Packaging Glass Bottle
Specific Gravity 1L = 1.69kg
Chemical Name or Material Phosphoric Acid
Grade Baker Analyzed™ ACS Reagent
Show More Show Less