Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, Diluted, 10% Solution, Multi-Compendial, N.F., J.T.Baker™

Click to view available options
Quantity:
19L
Packaging:
Poly Bottle
Poly Drum

Chemical Identifiers

7664-38-2
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
CHEBI:26078
Quantity 19L
CAS 7664-38-2
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES OP(=O)(O)O
IUPAC Name phosphoric acid
Molecular Weight (g/mol) 97.994
PubChem CID 1004
ChEBI CHEBI:26078
Formula Weight 98
Packaging Poly Bottle
Specific Gravity 1L = 1.05kg
Chemical Name or Material Phosphoric Acid, Diluted, 10% Solution
Grade Multi-Compendial/NF
Show More Show Less