Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, Multi-Compendial, NF, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
4L
Packaging:
Glass Bottle
Glass Bottle, Single ship
Poly Bottle
PVC Coated Glass Bottle
7664-38-2
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
orthophosphoric acid,sonac,o-phosphoric acid,phosphorsaeure,acidum phosphoricum,evits,wc-reiniger,acide phosphorique,white,acido fosforico
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
7664-38-2
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
orthophosphoric acid,sonac,o-phosphoric acid,phosphorsaeure,acidum phosphoricum,evits,wc-reiniger,acide phosphorique,white,acido fosforico
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
Quantity 2.5L
Formula Weight 98
Grade Multi-Compendial/NF
Packaging Glass Bottle
Specific Gravity 1L = 1.69kg
Chemical Name or Material Phosphoric Acid