Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
4L
19L
Packaging:
Glass Bottle
Glass Bottle, Mall
Glass Bottle, Single ship
Poly Bottle
Poly Drum

Chemical Identifiers

7664-38-2
H3O4P
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
Quantity 4L
Formula Weight 98
Packaging Poly Bottle
Specific Gravity 1L = 1.69kg
Chemical Name or Material Phosphoric Acid
Grade FCC/NF