Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric acid, pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
7664-38-2
H3O4P
97.994
MFCD00011340
NBIIXXVUZAFLBC-UHFFFAOYSA-N
orthophosphoric acid,sonac,o-phosphoric acid,phosphorsaeure,acidum phosphoricum,evits,wc-reiniger,acide phosphorique,white,acido fosforico
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
7664-38-2
H3O4P
97.994
MFCD00011340
NBIIXXVUZAFLBC-UHFFFAOYSA-N
orthophosphoric acid,sonac,o-phosphoric acid,phosphorsaeure,acidum phosphoricum,evits,wc-reiniger,acide phosphorique,white,acido fosforico
1004
CHEBI:26078
phosphoric acid
OP(=O)(O)O
Quantity 500g
Color White
Linear Formula H3PO4
Merck Index 15, 7456
Density 1.68
Formula Weight 98
Physical Form Crystals or Crystalline Powder
Percent Purity ≥98%
Grade Pure
Iron (Fe) 10ppm max.
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble ethanol
Sulfate 30ppm max.
Melting Point 42.0°C
Chemical Name or Material Phosphoric acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger