Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorous acid, 97%

Manufacturer:  Alfa Aesar 8940736

Quantity 500g

 View more versions of this product

Catalog No. AA8940736


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13598-36-2
82
UN2834
Phosphonic acid
H3PO3
MFCD00137258
14,7346
Phosphorous acid

Specifications

White
13598-36-2
Crystals
H3PO3
MFCD00137258
14,7346
Soluble in water and alcohol
Odorless
Phosphorous acid
500g
White crystals
97%
82
UN2834
Phosphonic acid
1.651g/cm3 at 20°C
Air sensitive
97%

Safety and Handling

SDS & Safety Documents

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHOSPHOROUS ACID

RTECSNumber : SZ6400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit