Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus, AAS Standard Solution, P 1000µg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 89887AP

Quantity 500mL

 View more versions of this product

Catalog No. AA89887AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7697-37-2,7723-14-0,7732-18-5
P in 5% HNO3
MFCD00133771
Phosphorus

Specifications

7697-37-2,7723-14-0,7732-18-5
P in 5% HNO3
Clear
Phosphorus
Clear liquid
MFCD00133771
500mL

Safety and Handling

SDS & Safety Documents

H314 : Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit