Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus oxybromide, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
7789-59-5
Br3OP
286.69
UXCDUFKZSUBXGM-UHFFFAOYSA-N
phosphorus oxybromide,phosphoric tribromide,phosphoryl bromide,phosphoryl tribromide,phosphorusoxybromide,phosphorus v oxybromide,unii-h98h8y87qq,phosphorousoxybromide,phosphoroyl tribromide,pobr3
24613
O=P(Br)(Br)Br
7789-59-5
Br3OP
286.69
UXCDUFKZSUBXGM-UHFFFAOYSA-N
phosphorus oxybromide,phosphoric tribromide,phosphoryl bromide,phosphoryl tribromide,phosphorusoxybromide,phosphorus v oxybromide,unii-h98h8y87qq,phosphorousoxybromide,phosphoroyl tribromide,pobr3
24613
O=P(Br)(Br)Br
C-X bond formation reagent
Assay Percent Range 95%
Merck Index 15,746
Formula Weight 286.69
Physical Form Crystals or Fused Mass
Percent Purity 95%
Packaging Glass bottle
Boiling Point 192.0°C
Color Brown-Gray to Colorless
Melting Point 56.0°C
Quantity 100g
Chemical Name or Material Phosphorus oxybromide
Show More Show Less

GHS H StatementCauses severe skin burns and eye damage.

GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF SWALLOWED: Rinse mouth.Do NOT induce vomiting.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.Immediately call a POISON CENTER or doctor/physician.

;GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.I

Recommended Storage : 4°C