Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus pentoxide, 98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
1314-56-3
O5P2
141.94
MFCD00011440
Phosphoric anhydride
1314-56-3
O5P2
141.94
MFCD00011440
Phosphoric anhydride
Adsorbent
Assay Percent Range 98%
Linear Formula P2O5
Fieser 01,871; 07,291; 10,319; 14,151; 15,267
Merck Index 15,7467
Solubility Information Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat
Formula Weight 141.94
Physical Form Hygroscopic Crystalline Powder
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Reducing Substances 0.1% max.
Solubility Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat.
Sublimation Point 0340.0°C
Color White
Melting Point 340.0°C to 360.0°C
Quantity 250g
Chemical Name or Material Phosphorus pentoxide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 215-236-1