Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus Standard, 1mL = 1mg P as Phosphate, 1000ppm P, Ricca Chemical
SDP

Catalog Number p-3252076
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural
7722-76-1, 7732-18-5
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
CHEBI:62982
phosphoric acid amine
N.OP(O)(O)=O
7722-76-1, 7732-18-5
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
CHEBI:62982
phosphoric acid amine
N.OP(O)(O)=O
Physical Form Liquid
Packaging Poly natural
Equivalence 1 mL = 1 mg P as Phosphate
Color Colorless
Quantity 120 mL
Concentration or Composition Notes 1000ppm P (3066ppm PO43-)
Chemical Name or Material Phosphorus Standard
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit